Learn More
TARGETMOL CHEMICALS INC ML335 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T422425MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ML335 is a selective activator of TREK-1 and TREK-2. Purity 99.73%
Catalog No. 50-287-9981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.