Learn More
Selleck Chemical LLC ML792 5mg
Supplier: Selleck Chemical LLC S86975MG
ML-792 is a potent and selective inhibitor ofSUMO (small ubiquitin-like modifier)-activating enzyme (SAE). ML-792 inhibits SAE/SUMO1 and SAE/SUMO2 in ATP¿inorganic pyrophosphate (PPi) exchange assays with IC50 of 3 nM and 11 nM, respectively. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.