Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters
GREENER_CHOICESDP

Catalog No. 22147609 Shop All VistaLab Technologies Inc. Products
Click to view available options
Volume (Metric):
5 μL
15 μL
30 μL
40 μL
60 μL
70 μL
75 μL
80 μL
90 μL
120 μL
400 μL
450 μL
600 μL
700 μL
800 μL
900 μL

Catalog No. 22-147-609

Supplier: VistaLab™ Technologies 1115

Only null left
Add to Cart
Add to Cart

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Capacity engraved on bonnet

Specifications

Accuracy ±1%
Autoclavable Not Autoclavable
Color Silver
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum Alloy, Stainless Steel
Model MLA Precision
Number of Channels 1
Sterility Non-sterile
Type Precision Pipetter
Volume (Metric) 600 μL
Includes Nozzle Insert, Calibration Key
Precision ±1%
Quantity 1 Ea.
Warranty Limited Lifetime
Length (English) 9 in.
Width (English) 2 in.
Height (English) 1.5 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.