Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters
GREENER_CHOICESDP

Catalog No. 22147610 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
5 μL
15 μL
30 μL
40 μL
60 μL
70 μL
75 μL
80 μL
90 μL
120 μL
400 μL
450 μL
600 μL
700 μL
800 μL
900 μL
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-147-610 800 μL
22-147-608 5 μL
22-147-613 15 μL
22-147-614 30 μL
22-147-615 40 μL
22-147-616 60 μL
22-147-617 70 μL
22-147-618 75 μL
22-147-619 80 μL
22-147-620 90 μL
22-147-621 120 μL
22-147-622 400 μL
22-147-609 600 μL
22-147-611 900 μL
22-147-612 450 μL
22-147-678 700 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22-147-610 Supplier VistaLab™ Technologies Supplier No. 1117
Add to Cart
Add to Cart

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.

  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Capacity engraved on bonnet

Specifications

Accuracy ±1%
Autoclavable Not Autoclavable
Color Silver
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum Alloy, Stainless Steel
Model MLA Precision
Number of Channels 1
Sterility Non-sterile
Type Precision Pipetter
Volume (Metric) 800 μL
Includes Nozzle Insert, Calibration Key
Precision ±0.75%
Quantity 1 Ea.
Warranty Limited Lifetime
Length (English) 9 in.
Width (English) 2 in.
Height (English) 1.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.