Learn More
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters

Shop All VistaLab Technologies Inc. Products


Description
- 'To deliver' capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Capacity engraved on bonnet
Specifications
Specifications
| Accuracy | ±2% |
| Autoclavable | Not Autoclavable |
| Color | Silver |
| Disposable | Reusable |
| For Use With | Micropipette Tip |
| Material | Aluminum Alloy, Stainless Steel |
| Model | MLA Precision |
| Number of Channels | 1 |
| Sterility | Non-sterile |
| Type | Precision Pipetter |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.