Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLANA Mouse anti-Human, Clone: UMAB217, liquid, UltraMAB™
SDP

Catalog No. 501685343
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5343 Supplier OriGene Supplier No. UM800109
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Specifications

Antigen MLANA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB217
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MLANA
Gene Accession No. Q16655
Gene Alias MART-1, MART1
Gene Symbols MLANA
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 48-118 of human MLANA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.