Learn More
Medchemexpress LLC HY-P1029 1mg Medchemexpress, MLCK inhibitor peptide 18 CAS:224579-74-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10291MG
Encompass_Preferred
Medchemexpress, HY-P1029 1mg MLCK inhibitor peptide 18 CAS:224579-74-2 MLCK inhibitor peptide 18 is a myosin light chain kinase ( MLCK ) inhibitor with an IC 50 of 50 nM, and inhibits CaM kinase II only at 4000-fold higher concentrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4138
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.