Learn More
Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, Natural Kynar, Cleanoom Packed, Male Slip Luer x 3/16" ID; 10/PK
Supplier:  Cole-Parmer 5011490
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. Natural Kynar fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. These robust fittings exhibit good mechanical properties, low moisture absorption, and good machinability.
Catalog No. 50-255-8012
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.