Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MLL5 Recombinant Protein Antigen
SDP

Catalog No. NB235894 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB235894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB235894 Supplier R&D Systems™ Supplier No. NBP317395PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 55904
Molecular Weight (g/mol) M.W. (Theoretical): 28 kDa
Quantity 0.1 mL
Storage Requirements Store at -20C. Avoid freeze-thaw cycles.
Gene Alias EC 2.1.1.43, FLJ10078, HDCMC04P, histone-lysine N-methyltransferase MLL5, KMT2EFLJ14026, Lysine N-methyltransferase 2E, MGC70452, myeloid/lymphoid or mixed-lineage leukemia 5 (trithorax homolog, Drosophila), Myeloid/lymphoid or mixed-lineage leukemia protein 5
Gene Symbol MLL5
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-17395It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.For current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here .
Research Category Cancer, Cell Biology, Cell Cycle and Replication, Stem Cells
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein MLL5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.