Learn More
Tocris Bioscience™ MLN 4924



Description
Specifications
Specifications
| Chemical Name or Material | [(1 S,2 S,4 R)-4-[4-[[(1 S)-2,3-Dihydro-1 H-inden-1-yl]amino]-7 H-pyrrolo[2,3-d]pyrimidin-7-yl]-2-hydroxycyclopentyl]methyl sulfamic acid ester |
| CAS | 905579-51-3 |
| Quantity | 50 mg |
| Target | NEDD8 Activating Enzyme E1) |
| Molecular Formula | C21H25N5O4S |
| Purity | 0.98 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.