Learn More
AdipoGen MLN4924
SDP
Supplier:  AdipoGen AGCR13703M025
Encompass_Preferred
Chemical. CAS 905579-51-3. Formula C21H25N5O4S . H2O. MW 443.5 . 18.0. Adenosine 5-monophosphate AMP analog. Potent, selective and cell permeable inhibitor of Nedd8 E1 Activating Enzyme NAE IC50=4.7nM. NEDD8 is an ubiquitin-like protein whose activity is required to activate cullin-RING ubiquitin E3 ligases CRLs responsible for the ubiquitination and proteasome-dependent turnover of certain substrates relevant to cancer cell survival. Anticancer agent with in vitro and in vivo activity. By inhibiting the Neddylation pathway this molecule disrupts cullin-RING ligase activity, leading to inhibition of proliferation or apoptotic death of various human tumor cells through the deregulation of S-phase DNA synthesis. Inhibits related enzymes ubiquitin-activating enzyme UAE and SUMO-activating enzyme SAE with IC50 values of 1.5 and 8.2µM, respectively. Angiogenesis inhibitor and autophagy inducer. Antiviral agent.
Catalog No. 50-256-2197
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.