Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLX Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5782 Supplier Bioss Supplier No. BS3950R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.

Specifications

Antigen MLX
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mlx
Gene Accession No. Q9UH92
Gene Alias BHLHD13; BigMax alpha; BigMax protein; BOS_19087; Class D basic helix-loop-helix protein 13; im:7146242; LOC100349784; MAD7; MAX dimerization protein MLX; Max-like bHLHZip protein; max-like factor protein; Max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; Tcfl4; TF4; transcription factor like 4; transcription factor-like 4; Transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409
Gene Symbols Mlx
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MLX.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6945
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.