Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MM 102
GREENER_CHOICESDP

Catalog No. 530710
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 53-071-0 Supplier Tocris Bioscience™ Supplier No. 5307/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WDR5/MLL interaction inhibitor

MM 102 is a potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM). Inhibits MLL1 methyltransferase activity and MLL-1-induced HoxA9 and Meis-1 gene expression in leukemia cells expressing the MLL1-AF9 fusion gene. Also inhibits cell growth and induces apoptosis in leukemia cells harbouring MLL1 fusion proteins.

Specifications

CAS 1883545-52-5
Quantity 10 mg
Product Type MM 102
Purity >98%
Molecular Weight (g/mol) 783.83

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.