Learn More
Medchemexpress LLC HY-100869A 1mg Medchemexpress, MM-589 (TFA) CAS:2097887-21-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100869A1MG
Encompass_Preferred
Medchemexpress, HY-100869A 1mg MM-589 (TFA) CAS:2097887-21-1 MM-589 TFA is a potent inhibitor of WD repeat domain 5 (WDR5) and mixed lineage leukemia (MLL) protein-protein interaction. MM-589 binds to WDR5 with an IC50 of 0.90 nM and inhibits the MLL H3K4 methyltransferase activity with an IC50 of 12.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7784