Learn More
World Precision Instrument MM-6 Micropositioner-505084
SDP
Supplier:  World Precision Instrument 505084
Encompass_Preferred
The MM-6 micropositioner is light weight, accurate, compact device designed to eliminate many research and design problems, especially in the case of space limitations. Hardened steel precision pre-loaded ball bearing slides and high resolution thumbscrew movement assure smooth motion, precision, and excellent load bearing capacity.
The black anodized aluminum body is made with a one-piece base and three piece construction. The top plate is designed around an M2 thread size on a 10 mm square hole pattern and central M4 tapped hole. The MM-6 is available in single, double, or triple stages. Adding stages is an easy job and requires no special tools. Triple stages come with an angle bracket to attach the Z-axis. The travel of the stage is 0.26" (6.5 mm) and the base is easily adapted to most surfaces.
Catalog No. 50-253-9258
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.