Learn More
TARGETMOL CHEMICALS INC MM3122 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T60085
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MM3122 (MM-3122) is a selective type II transmembrane serine protease (TMPRSS2) inhibitor IC50 = 0.34 nM. MM3122 effectively blocks TMPRSS2 thereby inhibiting the entry of SARS-CoV-2 and MERS-CoV into human cells. purity: 100%
Catalog No. 50-257-9678
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.