Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMAE Recombinant Rabbit Monoclonal Antibody (PSH0-03)
GREENER_CHOICE

Catalog No. FEMA550765 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550765 Supplier Invitrogen™ Supplier No. MA550765
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Product MA5-50765 was used in a Competitive ELISA and Indirect ELISA.

MMAE or monomethyl auristatin E, is a synthetic chemical compound that belongs to the class of tubulin inhibitors. It is commonly used as a payload in antibody-drug conjugates (ADCs) for targeted cancer therapy. MMAE functions by binding to tubulin, a protein involved in cell division, and disrupting the microtubule network necessary for cell division and growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMAE
Applications ELISA
Classification Recombinant Monoclonal
Clone PSH0-03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias Monomethyl auristatin E
Host Species Rabbit
Immunogen MMAE-OVA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical, Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Rabbit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.