Learn More
Invitrogen™ MMAE Recombinant Rabbit Monoclonal Antibody (PSH0-04)
Shop All Thermo Scientific Products

Description
Product MA5-50766 was used in a Competitive ELISA and Indirect ELISA.
Specifications
Specifications
| Antigen | MMAE |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | PSH0-04 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene Alias | Monomethyl auristatin E |
| Host Species | Rabbit |
| Immunogen | MMAE-OVA. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.