Learn More
MMD2 Polyclonal Antibody

Description
MMD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
Specifications
| Antigen | MMD2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | MMD2 |
| Gene Accession No. | Q8IY49, Q8R189 |
| Gene Alias | 4930518M15Rik; C88001; Mmd2; monocyte to macrophage differentiation associated 2; monocyte to macrophage differentiation factor 2; monocyte to macrophage differentiation-associated 2; monocyte-to-macrophage differentiation factor 2; monocyte-to-macrophage differentiation-associated protein 2; PAQR10; Progestin and adipoQ receptor family member 10; progestin and adipoQ receptor family member X |
| Gene Symbols | MMD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.