Learn More
Enzo Life Sciences MMP-1 proenzyme (human fibroblasts) (5µg)
SDP
Supplier:  Enzo Life Sciences ALX200418C005
Encompass_Preferred
Alternative name: Matrix metalloproteinase 1, Interstitial collagenase, Fibroblast collagenase. Concentration: ~80µg/ml (Pierce-BCA). Purity: ≥90% (SDS-PAGE, Western blot). MW: ~56kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.0, 300mM NaCl, 5mM CaCl2, 1µM ZnCl2, 0.05% BRIJ35 and 0.05% sodium azide. Source: Isolated from human rheumatoid synovial fibroblasts. Requires activation. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7873
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.