Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MMP-13 (Collagenase-3) Ab-1 Mouse Monoclonal Antibody with BSA and azide

Catalog No. p-4552778 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS825R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS825R1 Supplier Epredia™ Supplier No. MS825R1

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Mouse

Clone: VIIIA2

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Human recombinant collagenase-3 protein

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human chondrosarcoma SW1353 cells; Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation (Native verified)
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-13 (Collagenase-3) Ab-1
Applications Affinity Purification, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone VIIIA2
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Human recombinant collagenase-3 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.