Learn More
CPC Scientific DNP-Gly-Pro-Leu-Gly-Met-Arg-Gly-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMPS015B
Encompass
Highly sensitive and selective substrate for collagenase 3 (MMP-13) with a kcat/Km value of 4.22 x 106 M-1s-1 (PH= 7.5, T = 37°C)ONE-LETTER SEQUENCE: Dnp-GPLGMRGL-NH2MOLECULAR FORMULA: C40H64N14O12S1MOLECULAR WEIGHT:965.11STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1872435-02-3], SYNONYMS: Dnp-Gly-Pro-Leu-Gly-Met-Arg-Gly-Leu-NH2, Dnp-GPLGMRGL-NH2, Matrix Metalloproteinase-13 SubstrateRESEARCH AREA: CancerREFERENCES: S.J. Deng, et al., J. Biol. Chem., 275, 31422-31427 (2000)
Catalog No. 50-210-9778
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.