Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MMP-16/MT3-MMP Recombinant Protein Antigen
SDP

Catalog No. NBP86721PEP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP86721PEP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP86721PEP Supplier Novus Biologicals™ Supplier No. NBP186721PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human MMP16. The MMP-16/MT3-MMP Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 4325
Species Human
Purity >80% by SDS-PAGE and Coomassie blue staining
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) This Recombinant Antigen is Only Intended to be Used as a Blocking Agent to Confirm Antibody Specificity with The Corresponding Antibody, Catalog Number NBP2-53818. It is Purified by IMAC Chromatography, and The Expected Concentration is Greater Than 0.5 mg/ml
Gene Symbol MMP16
Molecular Weight (g/mol) 31 kDa
Preservative No Preservative
Quantity 100 μL
Source E. coli
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.