Learn More
ILEX LIFE SCIENCES LLC PromedeusLab Matrix Metalloproteinase 2 (MMP-2) Human ELISA Kit (96 wells)
SDP
Supplier:  ILEX LIFE SCIENCES LLC PL1023
Encompass_Preferred
PromedeusLab Matrix Metalloproteinase 2 (MMP-2) Human ELISA Kit (96 wells). DESCRIPTION: Enzyme immunoassay for the quantitative determination of matrix metalloproteinase 2 (MMP-2) in human serum and plasma samples. Quality controls are included with the kit. Developed and manufactured in Europe by PromedeusLab. SAMPLE TYPES: Serum, Plasma. ASSARY RANGE: 0.625 ng/mL - 40 ng/mL. SENSITIVITY: The limit of detection, defined as a concentration of human MMP-2 giving absorbance higher than absorbance of blank + 3 standard deviations, is better than 0.25 ng/mL of sample. OTHER NAMES: Matrix metalloproteinase-2 (MMP-2), MMP2, 72 kDa type IV collagenase, 72 kDa gelatinase, Gelatinase A, TBE-1, UniProtKB# P08253, EC:3.4.24.24.
Catalog No. 50-238-8556
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.