Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MMP-2/MMP-9 Inhibitor I, Solid, 99% Purity, 5 mg

Catalog No. 502705415
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5415 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5415 Supplier Abcam Supplier No. AB1451905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent matrix metalloproteinase inhibitor that synergizes with TNF-α to induce cancer cell apoptosis.

  • Purity: >99%
  • Molecular Weight: 381.4 Da
  • Form: Solid
  • Activity: Matrix metalloproteinase inhibitor of MMP-2/MMP-9
  • Additional Information: Synergizes with TNF-α to induce apoptosis in cancer cells. Active in vitro.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule MMP-2, MMP-9
Content And Storage Store at 4°C.
Buffer 100% MMP-2/MMP-9 Inhibitor I
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Quantity 5 mg
For Use With (Application) Cancer-apoptosis induction, In vitro enzyme inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.