Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-23 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6873 Supplier Bioss Supplier No. BS5875R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMP23B (matrix metallopeptidase 23B) is also named as MMP21, MMP22. It is a regulator in matrix remodeling and may be playing a role in enchondral bone formation and in regulation of cranial suture closure. It has 3 isoforms produced by alternative splicing with the molecular weight of 44 kDa, 48 kDa and 46 kDa, respectively. It also can be N-glycosylated.

Specifications

Antigen MMP-23
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MMP23B
Gene Accession No. O75900
Gene Alias Femalysin; matrix metallo protease; matrix metallopeptidase 23A (pseudogene); matrix metallopeptidase 23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; Matrix metalloproteinase-21; Matrix metalloproteinase-22; Matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR; MIFR-1; MMP; MMP 23; MMP21; MMP-21; MMP22; MMP-22; MMP23; MMP-23; MMP23A; MMP23B; MMPs; RP11-345P4.2
Gene Symbols MMP23B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MMP-23.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8510, 8511
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.