Learn More
Diagnostic Biosystems MMP-9 MMP-9
Supplier:  Diagnostic Biosystems RP06601
Encompass_Preferred
This antibody reacts with a 92 kDa pro form and 86 kDa active form of MMP9. MMPs are proteolytic enzymes capable of degrading connective tissue components. MMPs play a crucial role in tumor cell invasion and metastasis.
Catalog No. 50-238-5671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.