Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam MMP-9 Inhibitor, 511.6 Da, >95% Purity, 500 ug
Description
- Molecular Weight: 511.6 Da
- Purity: >95%
- Functional Role: Inhibitor of MMP-9, involved in tumor cell migration, invasion and angiogenesis
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | MMP-9 |
| Buffer | 100% MMP-9 inhibitor |
| Formulation | 100% MMP-9 Inhibitor |
| Quantity | 500 μg |
| For Use With (Application) | Tumor cell migration, tumor cell invasion, angiogenesis |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?