Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MMP Activity Assay Kit, Fluorometric, Red, 100/Test

Catalog No. 502584939
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4939 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4939 Supplier Abcam Supplier No. AB112147100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Validated assay kit for measuring MMP activity in cell and tissue extracts, with over 17 citations.

  • Assay Type: Direct
  • Detection Method: Fluorescent
  • Sample Types: Purified protein, Tissue Lysate
  • Applications: Enzyme activity assay
  • Cited in: > 15 publications
  • Compatible Equipment: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Matrix Metalloproteinase
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 100 Tests
Detection Location In-Solution Detection
Detection Method Fluorescence
Sample Type Purified protein, Tissue lysate
Quantity 100 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.