Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP1 Recombinant Mouse Monoclonal Antibody (K29034_8E3)

Catalog No. 780001M
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-000-1M 100 μL
78-000-1MXS 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 78-000-1M Supplier Invitrogen™ Supplier No. 780001M
Only null left
Add to Cart
Edge
Add to Cart

Mouse Recombinant Monoclonal Antibody

Recombinant mouse monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K29034_8E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene MMP1
Gene Accession No. P03956
Gene Alias 22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG; CLGN; Collagenase; collagenase-like A; fibroblast collagenase; Interstitial collagenase; interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1; matrix metallopeptidase 1 (interstitial collagenase); matrix metallopeptidase 1a; matrix metallopeptidase 1a (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; matrix metalloproteinase 1a (interstitial collagenase); Matrix metalloproteinase1; Matrix metalloproteinase-1; Matrix metalloproteinase-1a; McolA; Mcol-A; MMP; MMP1; MMP-1; Mmp1a; MMP-1a; MMPs
Gene Symbols MMP1
Host Species Mouse
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.