Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120848 Supplier Invitrogen™ Supplier No. PA5120848
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Raji, 293T, Mouse lung, Mouse kidney, Mouse skeletal muscle, Mouse ovary, Mouse heart. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: FRYPLSLSPD DCRGVQHLYG QPWPTVTSRT PALGPQAGID TNEIAPLEPD APPDACEASF DAVSTIRGEL FFFKAGFVWR LRGGQLQPGY PALASRHWQG LPSPVDAAFE DAQGHIWFFQ GAQYWVYDGE KPVLGPAPLT ELGLVRFPVH AALVWGPEKN KIYFFRGRDY WRFHPSTRRV DSPVPRRATD WRGVPSEIDA AFQDADGYAY FLRGRLYWKF DPVKVKALEG FPRLVGPDFF GCAEPANTFL.

Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mmp11
Gene Accession No. P24347, Q02853, Q499S5
Gene Alias matrix metallo protease; matrix metallopeptidase 11; matrix metallopeptidase 11 (stromelysin 3); Matrix metalloproteinase 11 (stromelysin 3); matrix metalloproteinase-11; MMP; MMP11; MMP-11; MMPs; SL-3; ST3; Stmy3; stromelysin III; stromelysin-3
Gene Symbols Mmp11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 239-488 of human MMP11 (NP_005931.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17385, 25481, 4320
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.