Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542893 Supplier Thermo Scientific Supplier No. PA542893
Only null left
Add to Cart
Add to Cart

MMP20 Polyclonal Antibody

MMP20 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP20
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MMP20
Gene Accession No. O60882
Gene Alias AI2A2; Enamel metalloproteinase; enamel metalloproteinase precursor; enamelysin; matrix metallo protease; matrix metallopeptidase 20; matrix metallopeptidase 20 (enamelysin); matrix metalloproteinase; matrix metalloproteinase 20; matrix metalloproteinase 20 (enamelysin); matrix metalloproteinase-20; MMP; MMP20; MMP-20; MMPs
Gene Symbols MMP20
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MMP20.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9313
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.