Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA125234
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA125234 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA125234 Supplier Invitrogen™ Supplier No. PA125234
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP21
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% sodium azide
Gene MMP21
Gene Accession No. O93470, Q8K3F2, Q8N119
Gene Alias b2b2458Clo; b2b873Clo; HTX7; matrix metallo protease; matrix metallopeptidase 21; matrix metallopeptidase 21 L homeolog; matrix metalloproteinase 21; matrix metalloproteinase-21; mmp; mmp21; MMP-21; mmp21.L; mmp21-A; MMPs; XELAEV_18034590mg; XMMP
Gene Symbols MMP21, mmp21.L
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the hinge region of human matrix metalloproteinase-21.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118856, 214766, 293573, 378572
Target Species Human, Mouse, Rat, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.