Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MMP8 ELISA Kit

Catalog No. EHMMP8
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHMMP8 96 Tests
EHMMP8X5 5 x 96 Tests
EHMMP8X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHMMP8 Supplier Invitrogen™ Supplier No. EHMMP8
Add to Cart
Add to Cart

ELISA

The Human Matrix MetalloProteinase 8 (Hu MMP8) ELISA quantitates Hu MMP8 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MMP8. Principle of the method The Human MMP8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Can degrade fibrillar type I, II, and III collagens.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22894
Assay Range 8.23 to 6000 pg/mL
Assay Sensitivity 6 pg/mL
Calibration Range 6 pg/mL to 6000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Cell Structure & Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the cytokines tested: human Angiopoietin-1, Angiostatin , B7-1, BMP-7, CD14, CD30, CD40, CD40 Ligand, CTLA-4, CXCL16, Dkk-4 , DR6, Endostatin, E-Selectin, Follistatin, HB-EGF, HVEM, ICAM-2, IGF-II, IL-10 Ra, IL-10 Rb, IL-18, IL-9 , IL-2 Ra, IL-2 Rb, IL-5 Ra, LAP, L-Selectin, M-CSF R, MMP-1, 2, 3, 7, 8, 9, 10 and 12, PDGF-AB, SDF-1 b, Tie-1, Tie-2, TIMP-3.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4317
Gene Symbol MMP8
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym CLG1, HNC, MMP-8, PMNL-CL
Target MMP8
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.