Learn More
TARGETMOL CHEMICALS INC MMRI64 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T280785MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MMRi64 is a specific inhibitor of Mdm2-MdmX E3 ligase activity.MMRi64 potently induces p53 accumulation and apoptosis in the p53 pathway in leukemia/lymphoma cells disrupts Mdm2-MdmX interactions and downregulates Mdm2 and MdmX in leukemia cells.MMRi64 MMRi64 can be used in cancer research. purity: 99%
Catalog No. 50-258-0551