Learn More
Medchemexpress LLC mmu-miR-155-5p mimic 5nmol | 5nmol
SDP
Supplier:  Medchemexpress LLC HYR026915NMOL
Encompass_Preferred
mmu-miR-155-5p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-21303