Learn More
Medchemexpress LLC Mn(III)TMPyP (pentachloride) | 125565-45-9 | MFCD01861931 | 95.0% | 909.01 g/mol | C44H36Cl5MnN8 | 5 MG

Supplier: Medchemexpress LLC HY1097485MG
Mn(III)TMPyP pentachloride is a redox-active manganese(III) porphyrin used as a biochemical reagent in life-science research. It acts as a superoxide dismutase (SOD) mimetic and peroxynitrite decomposition catalyst and is applied in studies of oxidative stress, cell signaling, and related in vitro assays. The material is supplied as a solid pentachloride salt with specified purity for experimental use.
- Functions as a superoxide dismutase (SOD) mimetic and peroxynitrite decomposition catalyst.
- Used in oxidative stress and cell signaling research and in vitro biochemical assays.
- Supplied as a solid pentachloride salt for stability and handling.
- Purity of 95.0% suitable for research applications.
- Molecular formula C44H36Cl5MnN8 and molecular weight 909.01 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.