Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101149 Supplier Invitrogen™ Supplier No. PA5101149
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MN1.

Meningioma 1 (MN1) contains two sets of CAG repeats. It is disrupted by a balanced translocation (4; 22) in a meningioma, and its inactivation may contribute to meningioma 32 pathogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MN1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MN1
Gene Accession No. Q10571
Gene Alias AA003644; AA009236; dJ353E16.2; meningioma (disrupted in balanced translocation) 1; meningioma (translocation balanced); meningioma 1; meningioma chromosome region 1; MGCR; MGCR1; MGCR1-PEN; MN1; MN1 proto-oncogene, transcriptional regulator; probable tumor suppressor protein MN1; RGD1565571; Transcriptional activator MN1
Gene Symbols MN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MN1(Accession Q10571), corresponding to amino acid residues L368-S418.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4330
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.