Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575319 Supplier Invitrogen™ Supplier No. PA575319
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Meningioma 1 (MN1) contains two sets of CAG repeats. It is disrupted by a balanced translocation (4; 22) in a meningioma, and its inactivation may contribute to meningioma 32 pathogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MN1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene MN1
Gene Accession No. D3YWE6, Q10571
Gene Alias AA003644; AA009236; dJ353E16.2; meningioma (disrupted in balanced translocation) 1; meningioma (translocation balanced); meningioma 1; meningioma chromosome region 1; MGCR; MGCR1; MGCR1-PEN; MN1; MN1 proto-oncogene, transcriptional regulator; probable tumor suppressor protein MN1; RGD1565571; Transcriptional activator MN1
Gene Symbols MN1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 830-880 of Human MN1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4330, 433938, 498194
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.