Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MNAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102517 Supplier Invitrogen™ Supplier No. PA5102517
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MNAT1.

Progression through the cell cycle requires activation of a series of enzymes designated cyclin dependent kinases (Cdks). The monomeric catalytic subunit, Cdk2, a critical enzyme for initiation of cell cycle progression, is completely inactive. Partial activation is achieved by the binding of regulatory cyclins such as cyclin D1, while full activation requires phosphorylation at Thr 160. The enzyme responsible for phosphorylation of Thr 160 in Cdk2 and also Thr 161 in Cdc2 p34, designated Cdk-activating kinase (CAK), has been partially purified and shown to be comprised of a catalytic subunit, a regulatory subunit and a subunit of unknown function. The regulatory subunit is a novel cyclin (cyclin H) and is required for activation of Cdk7. This previously undescribed protein, now termed Mat1, has been cloned as a protein that associates with the cyclin H-Cdk7 nuclear complex at all stages of the cell cycle. Cyclin H-Cdk7-Mat1 complexes display kinase activity towards Cdk activation domains, and the carboxy terminus of RNA polymerase II. Mat1 appears to constitute the first example of an assembly factor, essential for the formation of an active Cdk-cyclin complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNAT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mnat1
Gene Accession No. P51948, P51949
Gene Alias AdoMet synthetase; AMS1; CAP35; CDK7/cyclin H assembly factor; CDK7/cyclin-H assembly factor; CDK-activating kinase assembly factor MAT1; cyclin G1 interacting protein; cyclin-G1-interacting protein; E130115E11Rik; EC 2.5.1.6; Mat1; MATA1; MAT-I/III; Menage a trois; menage a trois 1; menage a trois 1 (CAK assembly factor); menage a trois homolog 1, cyclin H assembly factor; menage a trois-like protein 1 cyclin H assembly factor; MNAT CDK-activating kinase assembly factor 1; MNAT1; MNAT1, CDK activating kinase assembly factor; p35; p36; RING finger protein 66; RING finger protein MAT1; RNF66; TFB3
Gene Symbols Mnat1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MNAT1(Accession P51948), corresponding to amino acid residues H28-V78.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17420, 266713, 4331
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.