Learn More
Innovative Research Inc Mnky Rhss Ttl IgG Kit
Supplier:  Innovative Research Inc IMNRSIGGKTT
Encompass_Preferred
Monkey (Rhesus) Total IgG ELISA Kit - Monkey (Rhesus) Total IgG ELISA Kit from Innovative Research is intended for the quantitative determination of total IgG antigen in rhesus monkey serum, plasma, hybridoma cell supernatants, ascites or other biological fluids. The assay does not distinguish IgG subclasses. Strip well format. Reagents for up to 96 tests. Rhesus monkey IgG will bind to the monoclonal capture antibody coated on the microtiter plate. After appropriate washing steps, biotin labeled monoclonal anti-rhesus IgG primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody is reacted with horseradish peroxidase conjugated streptavidin. Following an additional washing step, TMB substrate is used for color development at 450nm.
Catalog No. 50-274-4218
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.