Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MNS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559016 Supplier Invitrogen™ Supplier No. PA559016
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKEEEENFRK TMLAKFAEDD RIELMNAQKQ RMKQLEHRRA VEKLIEERRQ QFLADKQREL EEWQLQQRRQ GFINAIIEEE RLKLLKEHAT NLLGYLPKGV FKKEDDIDLL GEEFRKVYQQ R Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

MNS1 may play a role in the control of meiotic division and germ cell differentiation through regulation of pairing and recombination during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MNS1
Gene Accession No. Q8NEH6
Gene Alias AW546487; meiosis specific nuclear structural 1; meiosis-specific nuclear structural 1; meiosis-specific nuclear structural protein 1; Mns1; SPATA40; spermatogenesis associated 40
Gene Symbols MNS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MNS1. Recombinant protein control fragment (Product #RP-98077).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55329
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.