Learn More
Apexbio Technology LLC NCT-501 1802088-50-1 5mg
SDP
Supplier:  Apexbio Technology LLC B60445
Encompass_Preferred
NCT-501 (CAS 1802088-50-1) is a theophylline-derived potent and selective inhibitor of aldehyde dehydrogenase 1A1 (ALDH1A1) ALDH1A1 plays a significant role in the metabolism of endogenous and exogenous aldehydes and is implicated in cancer biology particularly as a prognostic marker in prostate cancer and in cancer stem cell maintenance NCT-501 inhibits ALDH1A1 reversibly and has demonstrated inhibition of over 55% activity in a panel of more than 450 human kinases at 10 M In vivo intraperitoneal administration at 30 mg/kg yields higher plasma exposure and bioavailability compared to oral dosing NCT-501 serves as a valuable tool for investigating ALDH1A1 function and for studying aldehyde dehydrogenase involvement in cancer and metabolic disorders
Catalog No. 50-000-90171
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.