Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575591 Supplier Invitrogen™ Supplier No. PA575591
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

HCCA2 binds to and stimulates the kinase activity of the related human serine-threonine kinases NDR1 and NDR2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOB2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MOB2
Gene Accession No. Q70IA6, Q8VI63
Gene Alias 1110017M21Rik; 2700078K21Rik; AI256456; HCCA2; Mmh; MOB kinase activator 2; Mob2; Mob2 homolog; MOB2 Mps One Binder homolog; Mps one binder kinase activator-like 2; Ovary-specific MOB-like protein; RGD1562983
Gene Symbols MOB2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human MOB2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101513, 499288, 81532
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.