Learn More
Restek™ Mobi-Cap GL-45 Bottle Top Kits
SDP
Catalog No. 06611832 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Bottle Top Kit
Bottle Top Kit, Bottle (1 L)
Bottle Top Kit, Bottle (2 L)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
06-611-832 Bottle Top Kit
06-611-833 Bottle Top Kit, Bottle (1 L)
06-611-834 Bottle Top Kit, Bottle (2 L)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-611-832 Supplier Restek™ Supplier No. 27851
Add to Cart
Add to Cart
Convenient kits for easy mobile phase management
  • Protect mobile phases from evaporation and contamination, and laboratories from harmful vapors
  • Available with or without safety-coated GL-45 Bottle
  • Use one 1/4-28 threaded port for exhaust filter or air inlet valve
  • Other ports can be used as needed or closed with 1/4-28 plug (sold separately)
Specifications
Includes 3-Port Mobi-Cap GL-45 Bottles Top (1), PTFE Tubing (1/8 in. O.D. x 0.063 mm I.D. x 3 m), 1/16 in. Connection Sparger Filters (2): 2 μm/10 μm
Quantity 1 Kit
Thread Size (Metric) 78 mm
Neck Finish 78 mm
Product Type Bottle Top Kit