Learn More
Restek™ Mobi-Cap GL-45 Bottle Tops
SDP
Catalog No. 06611816 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Connector Cap (2-port)
Connector Cap (3-port)
Connector Cap (4-port)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
06-611-816 Connector Cap (2-port)
06-611-817 Connector Cap (3-port)
06-611-818 Connector Cap (4-port)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-611-816 Supplier Restek™ Supplier No. 27835
Add to Cart
Add to Cart
Protect mobile phases from evaporation and contamination while also protecting the lab from harmful vapors
  • Includes tubing connector nuts with color-sleeve adaptors and ferrules
  • Use one 1/4-28 threaded port for exhaust filter or air inlet valve
  • Other ports can be used as needed or closed with 1/4-28 plug (sold separately)
Specifications
Diameter (English) Holes: (1) 1/16 in. and (1) 1/8 in.
Includes Tubing Connector Nuts, Color-Sleeve Adaptors, and Ferrules
Material Polypropylene
Thread Size (Metric) 1/4-28
Neck Finish 78 mm
Product Type Connector Cap (2-port)
For Use With (Equipment) GL-45 Threaded Bottles