Description
- Includes tubing connector nuts with color-sleeve adaptors and ferrules
- Use one 1/4-28 threaded port for exhaust filter or air inlet valve
- Other ports can be used as needed or closed with 1/4-28 plug (sold separately)
Specifications
Specifications
| Diameter (English) | Holes: (1) 1/16 in. and (3) 1/8 in. |
| Includes | Tubing Connector Nuts, Color-Sleeve Adaptors, and Ferrules |
| Material | Polypropylene |
| Thread Size (Metric) | 1/4-28 |
| Neck Finish | 78 mm |
| For Use With (Equipment) | GL-45 Threaded Bottles |
| Product Type | Connector Cap (4-port) |