Learn More
Market Lab Marketlab Mobile Draw Station
Supplier:  Market Lab 10016
Encompass_Preferred
The Marketlab Mobile Draw Station is a mobile phlebotomy cart designed for blood collection and processing. It features a stainless steel work surface, storage for supplies, a sharps container, a wastebasket, and a glove box holder. The cart has 5" casters for easy mobility and can be customized with additional accessories like a computer arm, IV pole, and more. This phlebotomy cart provides a compact, efficient workspace to perform blood draws and specimen handling.
Catalog No. NC9755729
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.