Learn More
Market Lab Marketlab Mobile Draw Station Plus
Supplier:  Market Lab 42150
Encompass_Preferred
The Marketlab Mobile Draw Station Plus is a mobile phlebotomy cart designed for blood collection and processing. It features a stainless steel work surface, storage for supplies, a sharps container, a waste receptacle, locking casters, and a power strip. This cart allows phlebotomists to organize supplies and safely perform blood draws in various locations throughout a medical facility. Its mobility and ample storage make it useful for high-volume blood collection procedures.
Catalog No. NC1512636
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.