Learn More
Medicus Health Mobile Fixed Speed Centrifuge, 6 Place
SDP
Supplier:  Medicus Health 5260M1
Encompass
∙ Porta-Spin Portable Fixed Speed Centrifuge works on 12V car adapter This portable centrifuge is perfect for mobile ∙Phlebotomists and Traveling Nurses who need to collect and separate specimens ∙Mobile centrifuge is now designed with a clear lid ∙Designed for the professionals on the go∙Plugs into a car 12V power outlet∙ 6 place rotor for maximum tube size of 10ml tube 16 x 110 mm∙3300 RPM Fixed Speed for blood∙30 minute timer∙Dual lid latch for added safety∙Convenient stainless steel carrying handle∙Lid mounted “fish-eye” level to make leveling easy before spinning
Catalog No. NC1017909
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.